missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 306934-90-7
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD02677706
InChI Key CIZHAPZOOAJDCN-UHFFFAOYSA-N
Synonym 7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
PubChem CID 2776396
IUPAC Name 7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
SMILES C1COC2=C(C=C(C=C2)I)OC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC13211DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC13211DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC13211DE
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC13211DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

306934-90-7
276.073
CIZHAPZOOAJDCN-UHFFFAOYSA-N
2776396
C1COC2=C(C=C(C=C2)I)OC1
C9H9IO2
MFCD02677706
7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
7-iodo-3,4-dihydro-2H-1,5-benzodioxepine

Specifications

306934-90-7
100.0
Amber glass bottle
MFCD02677706
7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
C1COC2=C(C=C(C=C2)I)OC1
276.073
276.07
7-Iodo-3, 4-dihydro-2H-1, 5-benzodioxepine
94.5
95%
C9H9IO2
1g
CIZHAPZOOAJDCN-UHFFFAOYSA-N
7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
2776396
95%
SDS
missing translation for 'documents'

missing translation for 'documents'