missing translation for 'onlineSavingsMsg'
Learn More

7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC13211DA

 View more versions of this product

Catalog No. CC13211DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

7-Iodo-3, 4-dihydro-2H-1, 5-benzodioxepine
94.5
95%
C9H9IO2
1g
CIZHAPZOOAJDCN-UHFFFAOYSA-N
7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
2776396
95%
306934-90-7
100.0
Amber glass bottle
MFCD02677706
7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
C1COC2=C(C=C(C=C2)I)OC1
276.073
276.07

Chemical Identifiers

306934-90-7
276.073
CIZHAPZOOAJDCN-UHFFFAOYSA-N
2776396
C1COC2=C(C=C(C=C2)I)OC1
C9H9IO2
MFCD02677706
7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions