missing translation for 'onlineSavingsMsg'
Learn More

7-iodo-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific™

Chemical Identifiers

CAS 264617-03-0
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD11109311
InChI Key IASDAJCCAFGOCE-UHFFFAOYSA-N
Synonym 7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo
PubChem CID 33589431
IUPAC Name 7-iodo-2,3-dihydro-1-benzofuran
SMILES C1COC2=C1C=CC=C2I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC00911DA
SDS View Documents
Thermo Scientific Chemicals
CC00911DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC00911DE
SDS View Documents
Thermo Scientific Chemicals
CC00911DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

264617-03-0
246.047
IASDAJCCAFGOCE-UHFFFAOYSA-N
33589431
C1COC2=C1C=CC=C2I
C8H7IO
MFCD11109311
7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo
7-iodo-2,3-dihydro-1-benzofuran

Specifications

264617-03-0
100.0
Amber Glass Bottle
MFCD11109311
7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo
C1COC2=C1C=CC=C2I
246.047
246.04
7-Iodo-2, 3-dihydrobenzo[b]furan
96.5
97%
C8H7IO
1g
IASDAJCCAFGOCE-UHFFFAOYSA-N
7-iodo-2,3-dihydro-1-benzofuran
33589431
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'