missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-iodo-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC00911DA
Specifications
| 7-Iodo-2, 3-dihydrobenzo[b]furan | |
| 96.5 | |
| 97% | |
| C8H7IO | |
| 1g | |
| IASDAJCCAFGOCE-UHFFFAOYSA-N | |
| 7-iodo-2,3-dihydro-1-benzofuran | |
| 33589431 | |
| 97% |
| 264617-03-0 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD11109311 | |
| 7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo | |
| C1COC2=C1C=CC=C2I | |
| 246.047 | |
| 246.04 |
Chemical Identifiers
| 264617-03-0 | |
| 246.047 | |
| IASDAJCCAFGOCE-UHFFFAOYSA-N | |
| 33589431 | |
| C1COC2=C1C=CC=C2I |
| C8H7IO | |
| MFCD11109311 | |
| 7-iodo-2,3-dihydrobenzo b furan, 7-iodo-2,3-dihydrobenzofuran, benzofuran,2,3-dihydro-7-iodo | |
| 7-iodo-2,3-dihydro-1-benzofuran |