missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxythieno[3,2-b]pyridine 98.0+%, TCI America™

Catalog No. H16031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H16031G Supplier TCI America Supplier No. H16031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107818-20-2
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD01863636
InChI Key AACVULYSNJAKEQ-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8p
PubChem CID 5074765
IUPAC Name 4H-thieno[3,2-b]pyridin-7-one
SMILES C1=CNC2=C(C1=O)SC=C2

Specifications

Melting Point 235°C
Quantity 1 g
Formula Weight 151.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxythieno[3,2-b]pyridine