missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxycoumarin-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. H1352200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1352200MG 200 mg
H13521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1352200MG Supplier TCI America Supplier No. H1352200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 779-27-1
Molecular Formula C10H6O5
Molecular Weight (g/mol) 206.15
MDL Number MFCD00017491
InChI Key LKLWLDOUZJEHDY-UHFFFAOYSA-N
Synonym Umbelliferone-3-carboxylic Acid
PubChem CID 5337757
IUPAC Name 7-hydroxy-2-oxo-2H-chromene-3-carboxylic acid
SMILES OC(=O)C1=CC2=CC=C(O)C=C2OC1=O

Specifications

Melting Point 263°C
Color Yellow
Quantity 200 mg
Formula Weight 206.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxycoumarin-3-carboxylic Acid
TSCA No
Recommended Storage Freezer