accessibility menu, dialog, popup

UserName

CAS RN 779-27-1

IUPAC Name: 7-hydroxy-2-oxo-2H-chromene-3-carboxylic acid
Synonyms: 7-hydroxy-2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol): 206.15
Molecular Formula: C10H6O5
InChi Key: LKLWLDOUZJEHDY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=C(O)C=C2OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results