missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline, 98%, Thermo Scientific™

Chemical Identifiers

CAS 4602-73-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00143517
InChI Key UQBWYWCIBNWMPI-UHFFFAOYSA-N
Synonym 7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, pubchem12735, ksc496q9t, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline
PubChem CID 363043
IUPAC Name 6-methoxy-3,4-dihydroisoquinolin-7-ol
SMILES COC1=C(C=C2C=NCCC2=C1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC277822500
SDS View Documents
Thermo Scientific Chemicals
277822500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4602-73-7
177.2
UQBWYWCIBNWMPI-UHFFFAOYSA-N
363043
COC1=C(C=C2C=NCCC2=C1)O
C10H11NO2
MFCD00143517
7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, pubchem12735, ksc496q9t, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline
6-methoxy-3,4-dihydroisoquinolin-7-ol

Specifications

4602-73-7
100.0
Glass bottle
MFCD00143517
7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, pubchem12735, ksc496q9t, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline
UQBWYWCIBNWMPI-UHFFFAOYSA-N
6-methoxy-3,4-dihydroisoquinolin-7-ol
363043
98%
97.5
97.5% min. (GC)
C10H11NO2
250mg
Solubility in water: soluble in hot water. Other solubilities: soluble in chloroform,ethanol,methanol and hot,ethyl acetate
COC1=C(C=C2C=NCCC2=C1)O
177.2
177.2
7-Hydroxy-6-methoxy-3, 4-dihydroisoquinoline, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only