accessibility menu, dialog, popup

UserName

H3CONHO

CAS RN 4602-73-7

IUPAC Name: 6-methoxy-3,4-dihydroisoquinolin-7-ol
Synonyms: 6-methoxy-3,4-dihydroisoquinolin-7-ol
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: UQBWYWCIBNWMPI-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=NCC2)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)