missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-5-methyl-2-methylthio-[1,2,4]triazolo[1,5-a]pyrimidine 98.0+%, TCI America™

Catalog No. p-7130817
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09405G 5 g
H094025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09405G Supplier TCI America Supplier No. H09405G

Chemical Identifiers

CAS 40775-78-8
Molecular Formula C7H8N4OS
Molecular Weight (g/mol) 196.228
MDL Number MFCD00186154
InChI Key DITIBSDLGMCMTP-UHFFFAOYSA-N
Synonym 4-Hydroxy-6-methyl-2-methylmercapto-1,3,3a,7-tetraazaindene, 5-Methyl-2-methylthio-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 7-Hydroxy-5-methyl-2-methylthio-s-triazolo[1,5-a]pyrimidine
PubChem CID 101400
IUPAC Name 5-methyl-2-methylsulfanyl-1H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
SMILES CC1=CC(=O)N2C(=N1)N=C(N2)SC

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 196.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-5-methyl-2-methylthio-[1,2,4]triazolo[1,5-a]pyrimidine