accessibility menu, dialog, popup

UserName

CAS RN 40775-78-8

H3CSNNONHNCH3

CAS RN 40775-78-8

IUPAC Name: 5-methyl-2-(methylsulfanyl)-1H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
Synonyms: 5-methyl-2-(methylsulfanyl)-1H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
Molecular Weight (g/mol): 196.23
Molecular Formula: C7H8N4OS
InChi Key: DITIBSDLGMCMTP-UHFFFAOYSA-N
SMILES: CSC1=NC2=NC(C)=CC(=O)N2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results