missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4-(trifluoromethyl)coumarin 98.0+%, TCI America™

Catalog No. p-7131035
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14221G 1 g
H14225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14221G Supplier TCI America Supplier No. H14221G
Only null left

Chemical Identifiers

CAS 575-03-1
Molecular Formula C10H5F3O3
Molecular Weight (g/mol) 230.142
MDL Number MFCD00037578
InChI Key CCKWMCUOHJAVOL-UHFFFAOYSA-N
Synonym 7-hydroxy-4-trifluoromethyl coumarin, 7-hydroxy-4-trifluoromethylcoumarin, 7-hydroxy-4-trifluoromethyl-2h-chromen-2-one, 7-hydroxy-4-trifluoromethyl chromen-2-one, 4-trifluoromethyl umbelliferone, 7-hydroxy-4-trifluoromethyl-chromen-2-one, 2h-1-benzopyran-2-one, 7-hydroxy-4-trifluoromethyl, 7,4-hfc, maybridge1_006885, tfmu
PubChem CID 5375667
IUPAC Name 7-hydroxy-4-(trifluoromethyl)chromen-2-one
SMILES C1=CC2=C(C=C1O)OC(=O)C=C2C(F)(F)F

Specifications

Melting Point 185°C
Quantity 1 g
Formula Weight 230.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-4-(trifluoromethyl)coumarin