accessibility menu, dialog, popup

UserName

HOOOFFF

CAS RN 575-03-1

IUPAC Name: 7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one
Synonyms: 7-hydroxy-4-(trifluoromethyl)chromen-2-one
Molecular Weight (g/mol): 230.14
Molecular Formula: C10H5F3O3
InChi Key: CCKWMCUOHJAVOL-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)