missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 7-Hydroxy-4-methylcoumarin, 97%

Catalog No. p-146223
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156381000 100 g Plastic bottle
AC156385000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156381000 Supplier Thermo Scientific Chemicals Supplier No. 156381000
Only null left

CAS: 90-33-5 | C10H8O3 | 176.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-33-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00006866
InChI Key HSHNITRMYYLLCV-UHFFFAOYSA-N
Synonym 4-methylumbelliferone, hymecromone, 7-hydroxy-4-methylcoumarin, imecromone, 7-hydroxy-4-methyl-2h-chromen-2-one, cholestil, mendiaxon, 4-methyl-7-hydroxycoumarin, beta-methylumbelliferone, bilcolic
PubChem CID 5280567
ChEBI CHEBI:17224
IUPAC Name 7-hydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(O)=CC=C12

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 186°C to 191°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 18, 31
Merck Index 15, 4894
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in conc. h2so4,partly soluble in 5% aq. na2co3,partly soluble in ethanol and isopropanol
Formula Weight 176.17
Percent Purity 97%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-4-methylcoumarin, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
EINECSNumber 201-986-7
RTECSNumber GN7000000
TSCA TSCA

RUO – Research Use Only