missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4-methylcoumarin-3-acetic Acid 97.0+%, TCI America™

Catalog No. H1398100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1398100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1398100MG Supplier TCI America Supplier No. H1398100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5852-10-8
Molecular Formula C9H6N4O2
Molecular Weight (g/mol) 202.17
MDL Number MFCD00467589
InChI Key SGTFZCBIJHVSNT-UHFFFAOYSA-N
Synonym 4-Methylumbelliferone-3-acetic Acid
PubChem CID 5393149
IUPAC Name 2-(7-hydroxy-4-methyl-2-oxochromen-3-yl)acetic acid
SMILES CC1=C(C(=O)OC2=C1C=CC(=C2)O)CC(=O)O

Specifications

Melting Point 268°C
Color Yellow
Quantity 100 mg
Formula Weight 234.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-4-methylcoumarin-3-acetic Acid
TSCA No
Recommended Storage Freezer (-20 °C)