accessibility menu, dialog, popup

UserName

ONONNHN

CAS RN 5852-10-8

IUPAC Name: 2-(6-nitro-1H-1,3-benzodiazol-2-yl)acetonitrile
Synonyms: 2-(5-nitro-3H-1,3-benzodiazol-2-yl)acetonitrile
Molecular Weight (g/mol): 202.17
Molecular Formula: C9H6N4O2
InChi Key: SGTFZCBIJHVSNT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2N=C(CC#N)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results