missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-1-tetralone 98.0+%, TCI America™

Catalog No. p-7131142
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16501G 1 g
H16505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16501G Supplier TCI America Supplier No. H16501G
Only null left

Chemical Identifiers

CAS 22009-38-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD01312225
InChI Key LGFSAJZSDNYVCW-UHFFFAOYSA-N
Synonym 7-Hydroxy-3,4-dihydronaphthalen-1(2H)-one
PubChem CID 571818
IUPAC Name 7-hydroxy-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)O)C(=O)C1

Specifications

Melting Point 166°C
Boiling Point 215°C
Quantity 1 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxy-1-tetralone