missing translation for 'onlineSavingsMsg'
Learn More

7-(Fmoc-amino)heptanoic acid, 95%

Catalog No. p-7057012
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6628503 1 g
AAH6628506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6628503 Supplier Thermo Scientific Chemicals Supplier No. H6628503

CAS: 127582-76-7 | C22H25NO4 | 367.445 g/mol

7-(Fmoc-amino)heptanoic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-(Fmoc-amino)heptanoic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 127582-76-7
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.445
MDL Number MFCD01861353
InChI Key FRPYXTFBPCYALT-UHFFFAOYSA-N
Synonym fmoc-7-ahp-oh, 7-9h-fluoren-9-yl methoxy carbonyl amino heptanoic acid, 7-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, heptanoic acid, 7-9h-fluoren-9-ylmethoxy carbonyl amino, fmoc-ahp 7-oh, fmoc-nh-ch2 6-cooh, 7-fmoc-amino enanthic acid, 7-9h-fluoren-9-ylmethoxycarbonylamino heptanoic acid, fmoc-7-ahp-oh hplc, n-9-fluorenylmethoxycarbonyl-7-aminoenanthic acid
PubChem CID 2761803
IUPAC Name 7-(9H-fluoren-9-ylmethoxycarbonylamino)heptanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCC(=O)O

Specifications

Color White
Linear Formula FmocNH(CH2)6CO2H
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 367.44
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 7-(Fmoc-amino)heptanoic acid
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only