missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Ethynylcoumarin 98.0+%, TCI America™
Supplier: TCI America E10921G
Specifications
| 7-Ethynylcoumarin | |
| 134°C | |
| C11H6O2 | |
| 1 g | |
| O=C1OC2=CC(=CC=C2C=C1)C#C | |
| 170.17 | |
| 170.17 | |
| Crystalline Powder |
| 270088-04-5 | |
| White-Yellow | |
| MFCD28668960 | |
| NGKXZMHLZMPKFE-UHFFFAOYSA-N | |
| 7-ethynyl-2H-chromen-2-one | |
| 54110018 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 270088-04-5 | |
| 170.17 | |
| NGKXZMHLZMPKFE-UHFFFAOYSA-N | |
| 7-ethynyl-2H-chromen-2-one |
| C11H6O2 | |
| MFCD28668960 | |
| 54110018 | |
| O=C1OC2=CC(=CC=C2C=C1)C#C |
Safety and Handling
TSCA : No