accessibility menu, dialog, popup

UserName

HCOO

CAS RN 270088-04-5

IUPAC Name: 7-ethynyl-2H-chromen-2-one
Synonyms: 7-ethynylchromen-2-one
Molecular Weight (g/mol): 170.17
Molecular Formula: C11H6O2
InChi Key: NGKXZMHLZMPKFE-UHFFFAOYSA-N
SMILES: O=C1OC2=CC(=CC=C2C=C1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results