missing translation for 'onlineSavingsMsg'
Learn More

7-Ethynylcoumarin 98.0+%, TCI America™

Catalog No. E1092200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1092200MG 200 mg
E10921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1092200MG Supplier TCI America Supplier No. E1092200MG

Chemical Identifiers

CAS 270088-04-5
Molecular Formula C11H6O2
Molecular Weight (g/mol) 170.17
MDL Number MFCD28668960
InChI Key NGKXZMHLZMPKFE-UHFFFAOYSA-N
PubChem CID 54110018
IUPAC Name 7-ethynyl-2H-chromen-2-one
SMILES O=C1OC2=CC(=CC=C2C=C1)C#C

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 200 mg
Formula Weight 170.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Ethynylcoumarin