missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxycoumarin 97.0+%, TCI America™

Catalog No. E05385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05385G 5 g
E053825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05385G Supplier TCI America Supplier No. E05385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31005-02-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00006877
InChI Key LIFAQMGORKPVDH-UHFFFAOYSA-N
Synonym 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx
PubChem CID 35703
ChEBI CHEBI:28184
IUPAC Name 7-ethoxychromen-2-one
SMILES CCOC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 90°C
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Ethoxycoumarin