accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 31005-02-4

IUPAC Name: 7-ethoxy-2H-chromen-2-one
Synonyms: 7-ethoxycoumarin ethylumbelliferone
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: LIFAQMGORKPVDH-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2C=CC(=O)OC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)