Learn More
7-Ethoxycoumarin, 99%
CAS: 31005-02-4 | C11H10O3 | 190.198 g/mol
Supplier: Thermo Scientific Chemicals L14116ME
Description
7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.
Solubility
Soluble in hot methanol (very faint turbidity), 95% ethanol (50 mg/ml), DMF, and DMSO. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
| 7-Ethoxycoumarin | |
| 88°C to 91°C | |
| MFCD00006877 | |
| 146200 | |
| Soluble in hot methanol (very faint turbidity),95% ethanol (50mg/ml),DMF,and DMSO. Insoluble in water. | |
| CCOC1=CC2=C(C=C1)C=CC(=O)O2 | |
| 190.198 | |
| CHEBI:28184 | |
| 99% |
| 31005-02-4 | |
| C11H10O3 | |
| 500 mg | |
| 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx | |
| LIFAQMGORKPVDH-UHFFFAOYSA-N | |
| 7-ethoxychromen-2-one | |
| 35703 | |
| 190.2 |
Chemical Identifiers
| 31005-02-4 | |
| 190.198 | |
| LIFAQMGORKPVDH-UHFFFAOYSA-N | |
| 35703 | |
| 7-ethoxychromen-2-one |
| C11H10O3 | |
| MFCD00006877 | |
| 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx | |
| CHEBI:28184 | |
| CCOC1=CC2=C(C=C1)C=CC(=O)O2 |
Safety and Handling
EINECSNumber : 250-429-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only