missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxycoumarin, 99%

Catalog No. AAL14116MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14116MC 100 mg
AAL14116ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14116MC Supplier Thermo Scientific Chemicals Supplier No. L14116MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31005-02-4 | C11H10O3 | 190.198 g/mol

7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.

Solubility
Soluble in hot methanol (very faint turbidity), 95% ethanol (50 mg/ml), DMF, and DMSO. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 31005-02-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00006877
InChI Key LIFAQMGORKPVDH-UHFFFAOYSA-N
Synonym 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx
PubChem CID 35703
ChEBI CHEBI:28184
IUPAC Name 7-ethoxychromen-2-one
SMILES CCOC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 88°C to 91°C
Quantity 100 mg
Beilstein 146200
Solubility Information Soluble in hot methanol (very faint turbidity),95% ethanol (50mg/ml),DMF,and DMSO. Insoluble in water.
Formula Weight 190.2
Percent Purity 99%
Chemical Name or Material 7-Ethoxycoumarin
EINECSNumber 250-429-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only