missing translation for 'onlineSavingsMsg'
Learn More

7-(Dimethylamino)-4-methylcoumarin 98.0+%, TCI America™

Catalog No. p-7127687
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33555G 5 g
D335525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33555G Supplier TCI America Supplier No. D33555G
Only null left

Chemical Identifiers

CAS 87-01-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
InChI Key GZEYLLPOQRZUDF-UHFFFAOYSA-N
PubChem CID 6867
IUPAC Name 7-(dimethylamino)-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)N(C)C

Specifications

Melting Point 142°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Dimethylamino)-4-methylcoumarin
EINECSNumber (5)-3653
RTECSNumber GN6650000
TSCA Yes