accessibility menu, dialog, popup

UserName

CAS RN 87-01-4

IUPAC Name: 7-(dimethylamino)-4-methyl-2H-chromen-2-one
Synonyms: 7-(dimethylamino)-4-methylchromen-2-one
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: GZEYLLPOQRZUDF-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C2C(C)=CC(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results