missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Dimethylamino)-4-methylcoumarin 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 142°C |
| Quantity | 25 g |
| Formula Weight | 203.24 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 7-(Dimethylamino)-4-methylcoumarin |
Safety and Handling
| EINECSNumber | (5)-3653 |
| RTECSNumber | GN6650000 |
| TSCA | Yes |