missing translation for 'onlineSavingsMsg'
Learn More

7-(Diethylamino)coumarin-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. D4238100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4238100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4238100MG Supplier TCI America Supplier No. D4238100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50995-74-9
Molecular Formula C14H15NO4
Molecular Weight (g/mol) 261.277
MDL Number MFCD00068721
InChI Key WHCPTFFIERCDSB-UHFFFAOYSA-N
PubChem CID 659294
IUPAC Name 7-(diethylamino)-2-oxochromene-3-carboxylic acid
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)O

Specifications

Melting Point 229°C
Quantity 100 mg
Formula Weight 261.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)coumarin-3-carboxylic Acid