accessibility menu, dialog, popup

UserName

CAS RN 50995-74-9

IUPAC Name: 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylic acid
Synonyms: 7-(diethylamino)-2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol): 261.28
Molecular Formula: C14H15NO4
InChi Key: WHCPTFFIERCDSB-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C(O)=O)C(=O)OC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results