missing translation for 'onlineSavingsMsg'
Learn More

7-Diethylamino-4-methylcoumarin 98.0+%, TCI America™

Catalog No. p-7132272
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M063125G 25 g
M0631500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M063125G Supplier TCI America Supplier No. M063125G
Only null left

Chemical Identifiers

CAS 91-44-1
Molecular Formula C14H17NO2
Molecular Weight (g/mol) 231.295
MDL Number MFCD00006864
InChI Key AFYCEAFSNDLKSX-UHFFFAOYSA-N
Synonym 7-diethylamino-4-methylcoumarin, coumarin 1, 7-diethylamino-4-methyl-2h-chromen-2-one, coumarin 47, coumarin 460, 4-methyl-7-diethylamino coumarin, blancophor aw, blancophor ffg, 4-methyl-7-diethylaminocoumarin, hakkol p
PubChem CID 7050
ChEBI CHEBI:51938
IUPAC Name 7-(diethylamino)-4-methylchromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C

Specifications

Melting Point 72°C
Boiling Point 240°C
Quantity 25 g
Formula Weight 231.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Diethylamino-4-methylcoumarin
EINECSNumber (5)-2696
RTECSNumber GN6370000
TSCA Yes