accessibility menu, dialog, popup

UserName

H3CNCH3OOCH3

CAS RN 91-44-1

IUPAC Name: 7-(diethylamino)-4-methyl-2H-chromen-2-one
Synonyms: coumarin 1 diethylaminomethylcoumarin coumarin 47 coumarin 460
Molecular Weight (g/mol): 231.3
Molecular Formula: C14H17NO2
InChi Key: AFYCEAFSNDLKSX-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C(C)=CC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)