missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 7-Diethylamino-4-methylcoumarin, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 70°C to 73°C |
| Color | Beige to Purple |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Plastic bottle |
| Quantity | 100 g |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in aqueous acid solutions,soluble in nearly all common organic solvents,slightly soluble in aliphatic hydrocarbons |
| Formula Weight | 231.29 |
| Show More |
Safety and Handling
| EINECSNumber | 202-068-9 |
| RTECSNumber | GN6370000 |
| TSCA | TSCA |
RUO – Research Use Only