missing translation for 'onlineSavingsMsg'
Learn More

7-(Diethylamino)-3-(2-thienyl)coumarin 98.0+%, TCI America™

Catalog No. D50071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50071G 1 g
D50075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50071G Supplier TCI America Supplier No. D50071G

Chemical Identifiers

CAS 117850-52-9
Molecular Formula C17H17NO2S
Molecular Weight (g/mol) 299.39
MDL Number MFCD29089355
InChI Key ALJRTBKJTPXLCD-UHFFFAOYSA-N
PubChem CID 57494118
IUPAC Name 7-(diethylamino)-3-(thiophen-2-yl)-2H-chromen-2-one
SMILES CCN(CC)C1=CC=C2C=C(C3=CC=CS3)C(=O)OC2=C1

Specifications

Quantity 1 g
Formula Weight 299.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)-3-(2-thienyl)coumarin