accessibility menu, dialog, popup

UserName

H3CNCH3OOS

CAS RN 117850-52-9

IUPAC Name: 7-(diethylamino)-3-(thiophen-2-yl)-2H-chromen-2-one
Synonyms: 7-(diethylamino)-3-(thiophen-2-yl)chromen-2-one
Molecular Weight (g/mol): 299.39
Molecular Formula: C17H17NO2S
InChi Key: ALJRTBKJTPXLCD-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C3=CC=CS3)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results