missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 223°C |
| Quantity | 200 mg |
| Formula Weight | 347.42 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin |
Safety and Handling
| TSCA | Yes |