accessibility menu, dialog, popup

UserName

CAS RN 41044-12-6

H3CNCH3OONNH3C

CAS RN 41044-12-6

IUPAC Name: 7-(diethylamino)-3-(1-methyl-1H-1,3-benzodiazol-2-yl)-2H-chromen-2-one
Synonyms: 7-(diethylamino)-3-(1-methyl-1,3-benzodiazol-2-yl)chromen-2-one
Molecular Weight (g/mol): 347.42
Molecular Formula: C21H21N3O2
InChi Key: KZFUMWVJJNDGAU-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C3=NC4=CC=CC=C4N3C)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results