missing translation for 'onlineSavingsMsg'
Learn More

7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin 98.0+%, TCI America™

Catalog No. C28371G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28371G 1 g
C2837200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28371G Supplier TCI America Supplier No. C28371G
Only null left

Chemical Identifiers

CAS 41044-12-6
Molecular Formula C21H21N3O2
Molecular Weight (g/mol) 347.418
MDL Number MFCD00051349
InChI Key KZFUMWVJJNDGAU-UHFFFAOYSA-N
Synonym coumarin 30, 7-diethylamino-3-1-methyl-1h-benzo d imidazol-2-yl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-diethylamino-3-1-methyl-1h-benzimidazol-2-yl, ccris 4959, 7-diethylamino-3-1-methyl-1h-benzimidazol-2-yl-2h-chromen-2-one, 3-2-n-methylbenzimidazolyl-7-n,n-diethylaminocoumarin, 7-diethylamino-3-1-methylbenzimidazol-2-yl chromen-2-one, 7-diethylamino-3-1-methyl-1h-benzimidazol-2-yl-2-benzopyrone, 7-diethylamino-3-1-methyl-1,3-benzodiazol-2-yl chromen-2-one, coumarin 515
PubChem CID 100335
IUPAC Name 7-(diethylamino)-3-(1-methylbenzimidazol-2-yl)chromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4N3C

Specifications

Melting Point 223°C
Quantity 1 g
Formula Weight 347.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin