missing translation for 'onlineSavingsMsg'
Learn More

7-Deazapurine 98.0+%, TCI America™

Catalog No. D4820200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4820200MG 200 mg
D48201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4820200MG Supplier TCI America Supplier No. D4820200MG
Only null left

Chemical Identifiers

CAS 271-70-5
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD08234403
InChI Key JJTNLWSCFYERCK-UHFFFAOYSA-N
Synonym 7H-Pyrrolo[2,3-d]pyrimidine
PubChem CID 261591
IUPAC Name 7H-pyrrolo[2,3-d]pyrimidine
SMILES N1C=CC2=CN=CN=C12

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 200 mg
Formula Weight 119.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Deazapurine