accessibility menu, dialog, popup

UserName

HNNN

CAS RN 271-70-5

IUPAC Name: 7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: JJTNLWSCFYERCK-UHFFFAOYSA-N
SMILES: N1C=CC2=CN=CN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results