missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 7355-55-7 |
|---|---|
| Molecular Formula | C6H6N4O |
| Molecular Weight (g/mol) | 150.14 |
| InChI Key | OLAFFPNXVJANFR-UHFFFAOYSA-N |
| Synonym | 7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one |
| PubChem CID | 96253 |
| IUPAC Name | 2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one |
| SMILES | C1=CNC2=C1C(=O)N=C(N2)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC450260010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7355-55-7 | |
| 150.14 | |
| 7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one | |
| 2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one |
| C6H6N4O | |
| OLAFFPNXVJANFR-UHFFFAOYSA-N | |
| 96253 | |
| C1=CNC2=C1C(=O)N=C(N2)N |
Specifications
| 7355-55-7 | |
| C6H6N4O | |
| OLAFFPNXVJANFR-UHFFFAOYSA-N | |
| 2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one | |
| 96253 | |
| 90+% |
| 90% min. (HPLC) | |
| 7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one | |
| C1=CNC2=C1C(=O)N=C(N2)N | |
| 150.14 | |
| 150.14 | |
| 7-Deazaguanine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only