missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroisatin 98.0+%, TCI America™

Catalog No. C28405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28405G 5 g
C28401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28405G Supplier TCI America Supplier No. C28405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7477-63-6
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.575
MDL Number MFCD00022796
InChI Key MPLXQMMMGDYXIT-UHFFFAOYSA-N
Synonym 7-chloroisatin, 7-chloroindoline-2,3-dione, 7-chloro-2,3-dihydro-1h-indole-2,3-dione, 7-chloro-2,3-indolinedione, 1h-indole-2,3-dione, 7-chloro, 1h-indole-2,3-dione,7-chloro, 7-chloro-1h-benzo d azolidine-2,3-dione, 7-chlorisatin, 7-chloro isatin, pubchem15537
PubChem CID 344135
IUPAC Name 7-chloro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Cl)NC(=O)C2=O

Specifications

Melting Point 190°C
Quantity 5 g
Formula Weight 181.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloroisatin