accessibility menu, dialog, popup

UserName

ClOOHN

CAS RN 7477-63-6

IUPAC Name: 7-chloro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 7-chloro-1H-indole-2,3-dione
Molecular Weight (g/mol): 181.58
Molecular Formula: C8H4ClNO2
InChi Key: MPLXQMMMGDYXIT-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=C1NC(=O)C2=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)