missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroisatin, 97%

Catalog No. p-7052744
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5559403 1 g
AAH5559406 5 g
2 options

Catalog No. AAH5559403

Supplier: Thermo Scientific Chemicals H5559403

Only null left

CAS: 7477-63-6 | C8H4ClNO2 | 181.575 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7477-63-6
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.575
MDL Number MFCD00022796
InChI Key MPLXQMMMGDYXIT-UHFFFAOYSA-N
PubChem CID 344135
IUPAC Name 7-chloro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Cl)NC(=O)C2=O

Specifications

Melting Point 187°C to 191°C
Color Orange
Quantity 1 g
Synonym 7-chloroisatin, 7-chloroindoline-2,3-dione, 7-chloro-2,3-dihydro-1h-indole-2,3-dione, 7-chloro-2,3-indolinedione, 1h-indole-2,3-dione, 7-chloro, 1h-indole-2,3-dione,7-chloro, 7-chloro-1h-benzo d azolidine-2,3-dione, 7-chlorisatin, 7-chloro isatin, pubchem15537
Formula Weight 181.58
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Chloroisatin
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only