missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloroindole 98.0+%, TCI America™
$117.43 - $462.29
Chemical Identifiers
| CAS | 53924-05-3 |
|---|---|
| Molecular Formula | C8H6ClN |
| Molecular Weight (g/mol) | 151.593 |
| MDL Number | MFCD01321301 |
| InChI Key | WMYQAKANKREQLM-UHFFFAOYSA-N |
| Synonym | 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq |
| PubChem CID | 104644 |
| IUPAC Name | 7-chloro-1H-indole |
| SMILES | C1=CC2=C(C(=C1)Cl)NC=C2 |
Chemical Identifiers
| 53924-05-3 | |
| 151.593 | |
| WMYQAKANKREQLM-UHFFFAOYSA-N | |
| 104644 | |
| C1=CC2=C(C(=C1)Cl)NC=C2 |
| C8H6ClN | |
| MFCD01321301 | |
| 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq | |
| 7-chloro-1H-indole |
Specifications
| 53924-05-3 | |
| 95°C | |
| MFCD01321301 | |
| 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq | |
| C1=CC2=C(C(=C1)Cl)NC=C2 | |
| 151.593 | |
| 151.59 | |
| Crystalline Powder |
| 58°C | |
| C8H6ClN | |
| 1 g | |
| WMYQAKANKREQLM-UHFFFAOYSA-N | |
| 7-chloro-1H-indole | |
| 104644 | |
| ≥98.0% (GC) | |
| 7-Chloroindole |
Safety and Handling
TSCA : No