missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroindole 98.0+%, TCI America™

Catalog No. C24361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24361G 1 g
C24365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24361G Supplier TCI America Supplier No. C24361G

Chemical Identifiers

CAS 53924-05-3
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD01321301
InChI Key WMYQAKANKREQLM-UHFFFAOYSA-N
Synonym 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq
PubChem CID 104644
IUPAC Name 7-chloro-1H-indole
SMILES C1=CC2=C(C(=C1)Cl)NC=C2

Specifications

Melting Point 58°C
Boiling Point 95°C
Quantity 1 g
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloroindole