missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroindole, 98%

Catalog No. AAL17637MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17637MD 250 mg
AAL1763703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17637MD Supplier Thermo Scientific Chemicals Supplier No. L17637MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53924-05-3 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53924-05-3
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD01321301
InChI Key WMYQAKANKREQLM-UHFFFAOYSA-N
Synonym 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq
PubChem CID 104644
IUPAC Name 7-chloro-1H-indole
SMILES C1=CC2=C(C(=C1)Cl)NC=C2

Specifications

Melting Point 57°C to 58°C
Quantity 250 mg
Beilstein 114743
Formula Weight 151.6
Percent Purity 98%
Chemical Name or Material 7-Chloroindole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 258-865-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only