missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%

Catalog No. p-7032886
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3525603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3525603 Supplier Thermo Scientific Chemicals Supplier No. H3525603

CAS: 1020038-42-9 | C8H5ClN2O2 | 196.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1020038-42-9
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD09991784
InChI Key BEFUOITXMHRTRQ-UHFFFAOYSA-N
Synonym 7-chloroimidazo 1,2-a pyridine-2-carboxylic acid, 7-chloro-imidazo 1,2-a pyridine-2-carboxylic acid, 7-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
PubChem CID 28875810
IUPAC Name 7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CN2C=CC(Cl)=CC2=N1

Specifications

Melting Point ∼231°C (decomposition)
Quantity 1 g
Formula Weight 196.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 7-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only