accessibility menu, dialog, popup

UserName

CAS RN 1020038-42-9

IUPAC Name: 7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Synonyms: 7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 196.59
Molecular Formula: C8H5ClN2O2
InChi Key: BEFUOITXMHRTRQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN2C=CC(Cl)=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results