missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. C28975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28975G 5 g
C28971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28975G Supplier TCI America Supplier No. C28975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68077-26-9
Molecular Formula C12H8ClFNO3
Molecular Weight (g/mol) 268.65
MDL Number MFCD00792459
InChI Key WNNSMMJBBOPPOT-UHFFFAOYSA-M
Synonym 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-1-ethyl-6-fluoro-4-oxo-3-quinolinecarboxylic acid, 3-quinolinecarboxylic acid, 7-chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxo, 7-chloro-1-ethyl-6-fluoro-4-oxohydroquinoline-3-carboxylic acid, 7-chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid, 7-chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, 1-ethyl-7-chloro-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, zlchem 820
PubChem CID 483169
IUPAC Name 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
SMILES CCN1C=C(C([O-])=O)C(=O)C2=CC(F)=C(Cl)C=C12

Specifications

Melting Point 284°C
Color White-Yellow
Quantity 5 g
Formula Weight 269.66
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-1-ethyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid